ENA332672539 Enamine

5-fluoro-1-benzofuran-2-carboxylic acid

  • MDL number MFCD09038172
  • Cas Number : 89197-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 263...265
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74251

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-74251
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart