ENA332672543 Enamine

3-(chloromethyl)-1-(propan-2-yl)-1H-pyrazole

  • MDL number MFCD18262437
  • Cas Number : 1260659-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74576

Safety Information

signalword Danger
pcodes P264,P270,P280,P301+P312,P305+P351+P338,P310,P330,P501
MSDS https://www.enaminestore.com/catalog/EN300-74576
pictograms GHS05,GHS07
hstatements H302,H318
Availability details for
Quantity
  • Availability
  • Add to Cart