ENA332672545 Enamine

3-iodo-6-methylpyridazine

  • MDL number MFCD17016068
  • Cas Number : 1618-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78958

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78958
Availability details for
Quantity
  • Availability
  • Add to Cart