ENA332672548 Enamine

7-methyl-1-benzothiophene

  • MDL number MFCD00130098
  • Cas Number : 14315-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75116

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75116
Availability details for
Quantity
  • Availability
  • Add to Cart