ENA332672553 Enamine

3-(2-bromoethyl)-1,3-diazaspiro[4.4]nonane-2,4-dione

  • MDL number MFCD09815128
  • Cas Number : 150259-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79851

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79851
Availability details for
Quantity
  • Availability
  • Add to Cart