ENA332672554 Enamine

5-{[(4-chlorophenyl)methyl]amino}-2,3-dihydro-1H-1,3-benzodiazol-2-one

  • MDL number MFCD07610987
  • Cas Number : 878713-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79855

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79855
Availability details for
Quantity
  • Availability
  • Add to Cart