ENA332672568 Enamine

4-[(3-methoxyphenyl)amino]butanoic acid

  • MDL number MFCD09806780
  • Cas Number : 103565-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79880

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79880
Availability details for
Quantity
  • Availability
  • Add to Cart