ENA332672574 Enamine

6-chloro-1-methyl-2,3-dihydro-1H-indole-2-thione

  • MDL number MFCD19381939
  • Cas Number : 156136-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79106

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79106
Availability details for
Quantity
  • Availability
  • Add to Cart