ENA332672582 Enamine

1-(2-hydroxyethyl)piperidin-3-ol

  • MDL number MFCD14647094
  • Cas Number : 3554-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79118

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79118
Availability details for
Quantity
  • Availability
  • Add to Cart