ENA332672587 Enamine

N-(2-hydroxyethyl)octanamide

  • MDL number MFCD12180262

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79915
Availability details for
Quantity
  • Availability
  • Add to Cart