ENA332672593 Enamine

quinoline-6-carbonyl chloride hydrochloride

  • MDL number MFCD11501925
  • Cas Number : 158000-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79922
Availability details for
Quantity
  • Availability
  • Add to Cart