ENA332672594 Enamine

2-(cyclopropylamino)-1-(4-fluorophenyl)ethan-1-ol

  • MDL number MFCD11935486
  • Cas Number : 657419-26-6
  • Molecular Weight : 195.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-79923

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79923
Availability details for
Quantity
  • Availability
  • Add to Cart