ENA332672601 Enamine

2-amino-3-({[(4-methyl-1,3-thiazol-2-yl)carbamoyl]methyl}sulfanyl)propanoic acid

  • MDL number MFCD12411203

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79933

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79933
Availability details for
Quantity
  • Availability
  • Add to Cart