ENA332672608 Enamine

1-{2-[(4-methylpiperazin-1-yl)sulfonyl]phenyl}methanamine

  • MDL number MFCD09731561
  • Cas Number : 918865-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79209
Availability details for
Quantity
  • Availability
  • Add to Cart