ENA332672622 Enamine

2-(pyridin-3-yl)pyridine

  • MDL number MFCD00006215
  • Cas Number : 581-50-0
  • Molecular Weight : 156.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 30-32
Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-76175

Safety Information

signalword Danger
pcodes P201,P202,P261,P264,P270,P271,P280,P281,P301+P310,P302+P352,P304+P340,P305+P351+P338,P308+P313,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-76175
pictograms GHS06,GHS08
hstatements H301,H315,H319,H332,H335,H341,H351
Availability details for
Quantity
  • Availability
  • Add to Cart