ENA332672625 Enamine

(1R)-1-[3-(difluoromethoxy)phenyl]ethan-1-ol

  • MDL number MFCD18339576
  • Cas Number : 1315378-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79227

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79227
Availability details for
Quantity
  • Availability
  • Add to Cart