ENA332672634 Enamine

2,3-dihydro-1H-inden-2-ylmethanamine hydrochloride

  • MDL number MFCD07371495
  • Cas Number : 178955-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 262...264
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79989

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-79989
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart