ENA332672641 Enamine

3-hydroxypropanamide

  • MDL number MFCD04039564
  • Cas Number : 2651-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79996

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79996
Availability details for
Quantity
  • Availability
  • Add to Cart