ENA332672650 Enamine

4-(4-bromophenyl)-1,3-thiazole

  • MDL number MFCD02665938
  • Cas Number : 1826-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69...71
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79246

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79246
Availability details for
Quantity
  • Availability
  • Add to Cart