ENA332672664 Enamine

2-(5-ethylfuran-2-yl)piperidine

  • MDL number MFCD18652492
  • Cas Number : 1270404-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80021

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80021
Availability details for
Quantity
  • Availability
  • Add to Cart