ENA332672681 Enamine

6-(hydroxymethyl)piperidin-2-one

  • MDL number MFCD11110238
  • Cas Number : 174419-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80073

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80073
Availability details for
Quantity
  • Availability
  • Add to Cart