ENA332672690 Enamine

7-(hydroxymethyl)azepan-2-one

  • MDL number MFCD19220836
  • Cas Number : 150225-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95...97
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80082

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80082
Availability details for
Quantity
  • Availability
  • Add to Cart