ENA332672692 Enamine

4-chloroquinoline-3-carbonitrile

  • MDL number MFCD03093327
  • Cas Number : 69875-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128...130
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44043

Safety Information

msds https://www.enaminestore.com/catalog/EN300-44043
Availability details for
Quantity
  • Availability
  • Add to Cart