ENA332672695 Enamine

3-(2-methoxyethyl)-1,2-oxazol-5-amine

  • MDL number MFCD11102801
  • Cas Number : 1152534-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76572

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76572
Availability details for
Quantity
  • Availability
  • Add to Cart