ENA332672700 Enamine

2-(3-amino-1H-pyrazol-1-yl)-1-(morpholin-4-yl)ethan-1-one

  • MDL number MFCD12149354
  • Cas Number : 1179611-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80095

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80095
Availability details for
Quantity
  • Availability
  • Add to Cart