ENA332672701 Enamine

2-amino-1-[4-(difluoromethoxy)phenyl]ethan-1-ol

  • MDL number MFCD09040961
  • Cas Number : 926207-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54...56
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80096

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80096
Availability details for
Quantity
  • Availability
  • Add to Cart