ENA332672703 Enamine

cyclopentylmethanesulfonamide

  • MDL number MFCD14694939
  • Cas Number : 919354-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42...44
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80098

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80098
Availability details for
Quantity
  • Availability
  • Add to Cart