ENA332672705 Enamine

2-[2-(3-amino-1H-pyrazol-1-yl)ethoxy]ethan-1-ol

  • MDL number MFCD12785232
  • Cas Number : 1248122-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80100

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80100
Availability details for
Quantity
  • Availability
  • Add to Cart