ENA332672711 Enamine

1-chloro-2-(chloromethyl)-4-nitrobenzene

  • MDL number MFCD12025223
  • Cas Number : 69422-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61...63
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77118

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77118
Availability details for
Quantity
  • Availability
  • Add to Cart