ENA332672722 Enamine

6-chloro-N-(propan-2-yl)pyridazine-3-carboxamide

  • MDL number MFCD12165794
  • Cas Number : 345582-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80129

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80129
Availability details for
Quantity
  • Availability
  • Add to Cart