ENA332672744 Enamine

4-(cyclohexyloxy)benzoic acid

  • MDL number MFCD00053917
  • Cas Number : 139-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-188
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-80162

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80162
Availability details for
Quantity
  • Availability
  • Add to Cart