ENA332672757 Enamine

2-[2-(bromomethyl)-4-chlorophenoxy]acetonitrile

  • MDL number MFCD11180737
  • Cas Number : 1094373-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85...87
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80180

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80180
Availability details for
Quantity
  • Availability
  • Add to Cart