ENA332672759 Enamine

4-bromo-1-(4-fluorophenyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD12181673
  • Cas Number : 1156392-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80183

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80183
Availability details for
Quantity
  • Availability
  • Add to Cart