ENA332672780 Enamine

N-(cyanomethyl)-N-cyclopropylbenzamide

  • MDL number MFCD19348789
  • Cas Number : 1252203-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80208
Availability details for
Quantity
  • Availability
  • Add to Cart