ENA332672786 Enamine

3-(propan-2-yl)imidazo[1,5-a]pyridine-1-carbaldehyde

  • MDL number MFCD07381139
  • Cas Number : 1018516-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95...97
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79304

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79304
Availability details for
Quantity
  • Availability
  • Add to Cart