ENA332672788 Enamine

2-(2-phenylethoxy)ethan-1-amine hydrochloride

  • MDL number MFCD11506360
  • Cas Number : 1185298-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-80222

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80222
Availability details for
Quantity
  • Availability
  • Add to Cart