ENA332672789 Enamine

{6-bromo-1H-imidazo[4,5-b]pyridin-2-yl}methanol

  • MDL number MFCD19382196
  • Cas Number : 172648-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80223
Availability details for
Quantity
  • Availability
  • Add to Cart