ENA332672790 Enamine

2-(3-fluorophenyl)-2-hydroxypropanoic acid

  • MDL number MFCD14601994
  • Cas Number : 1083317-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80224
Availability details for
Quantity
  • Availability
  • Add to Cart