ENA332672793 Enamine

8-chloro-3,4-dihydro-2H-1,5-benzodioxepine-7-sulfonyl chloride

  • MDL number MFCD11104449
  • Cas Number : 1152581-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80231
Availability details for
Quantity
  • Availability
  • Add to Cart