ENA332672804 Enamine

5-amino-1-(2-hydroxyethyl)-1H-pyrazole-4-carboxamide

  • MDL number MFCD00090786
  • Cas Number : 58046-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182...184
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80245

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80245
Availability details for
Quantity
  • Availability
  • Add to Cart