ENA332672805 Enamine

5-(2-aminopropyl)-1-methyl-1H,4H,5H-pyrazolo[3,4-d]pyrimidin-4-one

  • MDL number MFCD09028370
  • Cas Number : 924871-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148...150
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80246

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80246
Availability details for
Quantity
  • Availability
  • Add to Cart