ENA332672825 Enamine

3-(4-fluorophenyl)propan-1-ol

  • MDL number MFCD09028733
  • Cas Number : 702-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-80278

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80278
Availability details for
Quantity
  • Availability
  • Add to Cart