ENA332672826 Enamine

1-[4-(pyridin-2-yl)piperazin-1-yl]propan-2-amine

  • MDL number MFCD09805457
  • Cas Number : 337956-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80279

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80279
Availability details for
Quantity
  • Availability
  • Add to Cart