ENA332672827 Enamine

N-[(4-aminophenyl)methyl]methanesulfonamide

  • MDL number MFCD01310487
  • Cas Number : 81880-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-80282

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80282
Availability details for
Quantity
  • Availability
  • Add to Cart