ENA332672832 Enamine

1-(2,3-dichlorophenyl)ethan-1-amine

  • MDL number MFCD06654499
  • Cas Number : 39226-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80287

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80287
Availability details for
Quantity
  • Availability
  • Add to Cart