ENA332672839 Enamine

3-cyclohexylprop-2-enoic acid

  • Cas Number : 4484-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80296
Availability details for
Quantity
  • Availability
  • Add to Cart