ENA332672857 Enamine

1-[3-(1H-1,2,4-triazol-1-yl)phenyl]methanamine

  • MDL number MFCD08271898
  • Cas Number : 741717-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80327

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80327
Availability details for
Quantity
  • Availability
  • Add to Cart