ENA332672858 Enamine

2-(3-methylphenyl)azepane

  • MDL number MFCD02663746
  • Cas Number : 383129-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80328

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80328
Availability details for
Quantity
  • Availability
  • Add to Cart