ENA332672869 Enamine

5'-bromo-1',2'-dihydrospiro[cyclopentane-1,3'-indole]-2'-one

  • MDL number MFCD13192266
  • Cas Number : 304876-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79336

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79336
Availability details for
Quantity
  • Availability
  • Add to Cart