ENA332672870 Enamine

(3-chloropropoxy)benzene

  • MDL number MFCD03940223

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 13-Dec
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80349

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80349
Availability details for
Quantity
  • Availability
  • Add to Cart